Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trifluorobenzyl Cyanide 97.0+%, TCI America™
SDP

Catalog No. T24145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T24145G Supplier TCI America Supplier No. T24145G

Chemical Identifiers

CAS 220141-74-2
Molecular Formula C8H4F3N
Molecular Weight (g/mol) 171.12
MDL Number MFCD00061220
InChI Key JTYBTJVFXUKNKW-UHFFFAOYSA-N
Synonym 2,4,5-trifluorobenzyl cyanide, 2,4,5-trifluorophenylacetonitrile, 2-2,4,5-trifluorophenyl acetonitrile, 2,4,5-trifluoro-phenyl-acetonitrile, 2-2,4,5-trifluorophenyl ethanenitrile, pubchem4206, acmc-209fqp, 2,4,5-trifluorobenzylcyanide, ksc494k1d, attercop-chm at118419
PubChem CID 2777957
IUPAC Name 2-(2,4,5-trifluorophenyl)acetonitrile
SMILES FC1=CC(F)=C(CC#N)C=C1F

Specifications

Color Yellow
Boiling Point 78°C
Quantity 5 g
UN Number 3276
Formula Weight 171.12
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4,5-Trifluorobenzyl Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.