accessibility menu, dialog, popup

UserName

Viewing profile as user:
OBrNOOCl

CAS RN 22019-49-4

IUPAC Name: 2-bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one
Synonyms: 2-bromo-1-(4-chloro-3-nitrophenyl)ethanone
Molecular Weight (g/mol): 278.49
Molecular Formula: C8H5BrClNO3
InChi Key: FXNQTMRPLLNPQY-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C=CC(=C1)C(=O)CBr

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results