Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one, Technical Grade, Thermo Scientific™

Catalog No. JP00700DA
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
JP00700DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. JP00700DA Supplier Thermo Scientific Chemicals Supplier No. JP00700DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22019-49-4
Molecular Formula C8H5BrClNO3
Molecular Weight (g/mol) 278.486
InChI Key FXNQTMRPLLNPQY-UHFFFAOYSA-N
PubChem CID 2778330
IUPAC Name 2-bromo-1-(4-chloro-3-nitrophenyl)ethanone
SMILES C1=CC(=C(C=C1C(=O)CBr)[N+](=O)[O-])Cl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 2-bromo-1-4-chloro-3-nitrophenyl ethanone, 2-bromo-1-4-chloro-3-nitrophenyl ethan-1-one, 4-chloro-3-nitrophenacylbromide, 4-chloro-3-nitrophenacyl bromide, ethanone,2-bromo-1-4-chloro-3-nitrophenyl, 4-chloro-3-nitro-phenacyl bromide, 4-chloro-3-nitrophen-acyl bromide, 2-bromo-4'-chloro-3'-nitroacetophenone, 2-bromo-1-4-chloro-3-nitro-phenyl ethanone, ethanone, 2-bromo-1-4-chloro-3-nitrophenyl
Formula Weight 278.49
Percent Purity 95%
Chemical Name or Material 2-Bromo-1-(4-chloro-3-nitrophenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.