accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 220204-00-2

IUPAC Name: [3'-(dihydroxyboranyl)-2,2'-dimethoxy-[1,1'-binaphthalen]-3-yl]boronic acid
Synonyms: 3'-(dihydroxyboranyl)-2,2'-dimethoxy-[1,1'-binaphthalen]-3-ylboronic acid
Molecular Weight (g/mol): 402.02
Molecular Formula: C22H20B2O6
InChi Key: PNKNNZOHEMBDRL-UHFFFAOYSA-N
SMILES: COC1=C(C=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC(B(O)O)=C1OC)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results