accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHOFFFOH

CAS RN 220239-68-9

IUPAC Name: 4-hydroxy-3-(trifluoromethyl)benzoic acid
Synonyms: 4-hydroxy-3-(trifluoromethyl)benzoic acid
Molecular Weight (g/mol): 206.12
Molecular Formula: C8H5F3O3
InChi Key: DPVRVZQEDJVWLS-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=C(O)C=C1)C(F)(F)F
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results