Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Hydroxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Catalog No. AC449122500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449122500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC449122500 Supplier Thermo Scientific Chemicals Supplier No. 449122500

Chemical Identifiers

CAS 220239-68-9
Molecular Formula C8H5F3O3
Molecular Weight (g/mol) 206.12
MDL Number MFCD01091008
InChI Key DPVRVZQEDJVWLS-UHFFFAOYSA-N
Synonym 4-hydroxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzoicacid, acmc-1cdpd, ksc495o6p, 4-hydroxy-3-trifluoromethyl-benzoic acid, 4-oxidanyl-3-trifluoromethyl benzoic acid
PubChem CID 2775112
IUPAC Name 4-hydroxy-3-(trifluoromethyl)benzoic acid
SMILES OC(=O)C1=CC(=C(O)C=C1)C(F)(F)F

Specifications

Melting Point 144.0°C to 147.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CF3C6H3CO2H(OH)
Quantity 250 mg
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzoic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.