accessibility menu, dialog, popup

UserName

Viewing profile as user:
FBrFFF

CAS RN 220239-69-0

IUPAC Name: 2-(bromomethyl)-1-fluoro-4-(trifluoromethyl)benzene
Synonyms: 2-(bromomethyl)-1-fluoro-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 257.03
Molecular Formula: C8H5BrF4
InChi Key: BYRMZRWUEAESEZ-UHFFFAOYSA-N
SMILES: FC1=CC=C(C=C1CBr)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results