Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 2-Fluoro-5-(trifluoromethyl)benzyl bromide

Catalog No. CDS003078 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS003078 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS003078 Supplier Sigma Aldrich Supplier No. CDS003078250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-hexene can be used:
  • As a substrate for polymerization by various catalysts
  • To prepare good quality middle distillates and lubricating oils by oligomerization
  • In the synthesis of aldehydes using supercritical co2 and rhodium catalyst by hydroformylation reaction

Specifications

CAS 220239-69-0
Molecular Formula C8H5BrF4
Linear Formula C8H5BrF4
MDL Number MFCD00061174
Quantity 1 L
Molecular Weight (g/mol) 257.02
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.