accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHN(R)(S)

CAS RN 22031-52-3

IUPAC Name: (1S,5R)-6-azabicyclo[3.2.0]heptan-7-one
Synonyms: (1S,5R)-6-azabicyclo[3.2.0]heptan-7-one
Molecular Weight (g/mol): 111.14
Molecular Formula: C6H9NO
InChi Key: AXDBIGNYXNSBHV-CRCLSJGQSA-N
SMILES: O=C1N[C@@H]2CCC[C@H]12
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results