Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

6-Azabicyclo[3.2.0]heptan-7-one, 98%, Thermo Scientific™

Catalog No. AC335835000
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC335835000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC335835000 Supplier Thermo Scientific Chemicals Supplier No. 335835000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22031-52-3
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD01863277
InChI Key AXDBIGNYXNSBHV-CRCLSJGQSA-N
Synonym 6-azabicyclo 3.2.0 heptan-7-one, 6-azabicyclo 3,2,0 heptan-7-one, 6-azabicyclo 3.2.0 heptane-7-one, 6-azabicyclo 3.2.0 heptan-7-one #
PubChem CID 549320
IUPAC Name 6-azabicyclo[3.2.0]heptan-7-one
SMILES O=C1N[C@@H]2CCC[C@H]12

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 45.0°C to 50.0°C
Color White to Yellow
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 500 mg
Solubility Information Solubility in water: .. Other solubilities: soluble in ether, chloroform, methanol
Formula Weight 111.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Azabicyclo[3.2.0]heptan-7-one, 98%
Show More Show Less
Hazard Category LIGHT SENSITIVE

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.