CAS RN 220465-43-0
CAS RN 220465-43-0
IUPAC Name:
(1H-indol-4-yl)boronic acid
Synonyms:
1H-indol-4-ylboronic acid
Molecular Weight (g/mol):
160.97
Molecular Formula:
C8H8BNO2
InChi Key:
USJUQEVUEBCLLR-UHFFFAOYSA-N
SMILES:
OB(O)C1=C2C=CNC2=CC=C1
1
–
1
of
1
results
1
Promotions Available
Sigma Aldrich indole-4-boronic acid
Sigma Aldrich indole-4-boronic acid
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.