CAS RN 22065-85-6
IUPAC Name:
1-benzylpiperidine-4-carbaldehyde
Synonyms:
1-benzylpiperidine-4-carbaldehyde
Molecular Weight (g/mol):
203.29
Molecular Formula:
C13H17NO
InChi Key:
SGIBOXBBPQRZDM-UHFFFAOYSA-N
SMILES:
O=CC1CCN(CC2=CC=CC=C2)CC1
1
–
1
of
1
results
1
1-Benzyl-4-piperidinecarboxaldehyde 96.0+%, TCI America™
CAS: 22065-85-6 Molecular Formula: C13H17NO Molecular Weight (g/mol): 203.285 MDL Number: MFCD02258902 InChI Key: SGIBOXBBPQRZDM-UHFFFAOYSA-N Synonym: 1-benzyl-4-piperidinecarboxaldehyde,n-benzylpiperidine-4-carboxaldehyde,1-benzyl-4-formylpiperidine,1-benzylpiperidine-4-carboxaldehyde,1-benzy-4-piperidine carboxyaldehyde,1-benzyl-4-piperidinecarbaldehyde,1-benzyl-piperidine-4-carbaldehyde,n-benzyl piperidine-4-carboxaldehyde,4-piperidinecarboxaldehyde, 1-phenylmethyl,n-benzyl-4-formylpiperidine PubChem CID: 89584 IUPAC Name: 1-benzylpiperidine-4-carbaldehyde SMILES: C1CN(CCC1C=O)CC2=CC=CC=C2