Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-4-piperidinecarboxaldehyde 96.0+%, TCI America™
SDP

Catalog No. B31905G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31905G 5 g
B319025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31905G Supplier TCI America Supplier No. B31905G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22065-85-6
Molecular Formula C13H17NO
Molecular Weight (g/mol) 203.285
MDL Number MFCD02258902
InChI Key SGIBOXBBPQRZDM-UHFFFAOYSA-N
Synonym 1-benzyl-4-piperidinecarboxaldehyde, n-benzylpiperidine-4-carboxaldehyde, 1-benzyl-4-formylpiperidine, 1-benzylpiperidine-4-carboxaldehyde, 1-benzy-4-piperidine carboxyaldehyde, 1-benzyl-4-piperidinecarbaldehyde, 1-benzyl-piperidine-4-carbaldehyde, n-benzyl piperidine-4-carboxaldehyde, 4-piperidinecarboxaldehyde, 1-phenylmethyl, n-benzyl-4-formylpiperidine
PubChem CID 89584
IUPAC Name 1-benzylpiperidine-4-carbaldehyde
SMILES C1CN(CCC1C=O)CC2=CC=CC=C2

Specifications

Boiling Point 128°C
Quantity 5 g
Formula Weight 203.29
Percent Purity ≥96.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-Benzyl-4-piperidinecarboxaldehyde
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.