accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22072-19-1

IUPAC Name: thian-3-ol
Synonyms: thian-3-ol
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: ZHHOMIAGFRSWCU-UHFFFAOYNA-N
SMILES: OC1CCCSC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results