Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrahydrothiopyran-3-ol 95%, Thermo Scientific™

Catalog No. AC166930250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC166930250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC166930250 Supplier Thermo Scientific Chemicals Supplier No. 166930250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22072-19-1
Molecular Formula C5H10OS
Molecular Weight (g/mol) 118.194
InChI Key ZHHOMIAGFRSWCU-UHFFFAOYSA-N
Synonym tetrahydrothiopyran-3-ol, 3-hydroxytetrahydrothiopyran, 2h-thiopyran-3-ol, tetrahydro, tetrahydro-2h-thiopyran-3-ol, 3-hydroxythianaphthen, thiacyclohexane-3-ol, tetrahyrothiopyran-3-ol, 3-thia-1-cyclohexanol, tetrahydrothiapyran-3-ol, thiopyran-3-ol, tetrahydro
PubChem CID 90725
IUPAC Name thian-3-ol
SMILES C1CC(CSC1)O

Specifications

Color Colorless
Density 1.14
Infrared Spectrum Authentic
Refractive Index 1.5341 to 1.5361 (20°C, 589nm)
Quantity 25 g
Beilstein 17, V,3, 110
Formula Weight 150.13
Percent Purity ≥94%
Physical Form Viscous Liquid
Chemical Name or Material Tetrahydrothiopyran-3-ol, 95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.