accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClCH3OCH3HN

CAS RN 2208-07-3

IUPAC Name: hydrogen ethyl ethanimidate chloride
Synonyms: hydrogen ethyl ethanimidate chloride
Molecular Weight (g/mol): 123.58
Molecular Formula: C4H10ClNO
InChi Key: WGMHMVLZFAJNOT-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CCOC(C)=N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results