Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

trans-1,2-Dimethylcyclohexane, MP Biomedicals

Catalog No. ICN205128 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN205128 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN205128 Supplier MP Biomedicals, Inc Supplier No. 0520512805

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2208-07-3
Molecular Formula C4H10ClNO
Molecular Weight (g/mol) 123.58
InChI Key WGMHMVLZFAJNOT-UHFFFAOYSA-N
Synonym ethyl acetimidate hydrochloride, ethyl ethanimidate hydrochloride, ethanimidic acid, ethyl ester, hydrochloride, ethyl ethanimidoate hydrochloride, ethylacetimidatehydrochloride, ethyl iminoacetate hydrochloride, ea hydrochloride, acetimidic acid, hydrochloride, ethanimidic acid, ethyl ester, hydrochloride 1:1, ethanimidic acid, hydrochloride
PubChem CID 2724290
IUPAC Name ethyl ethanimidate;hydrochloride
SMILES CCOC(=N)C.Cl

Specifications

Quantity 10 g
Formula Weight 112.22
Chemical Name or Material trans-1, 2-Dimethylcyclohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.