accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22098-10-8

IUPAC Name: ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
Synonyms: ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
Molecular Weight (g/mol): 263.11
Molecular Formula: C8H7BrO3S
InChi Key: PMBGHMBDWGNJJE-UHFFFAOYSA-N
SMILES: CCOC(=O)C(=O)C1=CC=C(Br)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results