Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ethyle2-(5-bromo-2-thienyl)-2-oxoacetate, 97%, Thermo Scientific™

Catalog No. BTB09133EA
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BTB09133EA 10 g
BTB09133DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BTB09133EA Supplier Thermo Scientific Chemicals Supplier No. BTB09133EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22098-10-8
Molecular Formula C8H7BrO3S
Molecular Weight (g/mol) 263.105
MDL Number MFCD00085056
InChI Key PMBGHMBDWGNJJE-UHFFFAOYSA-N
Synonym ethyl 2-5-bromothiophen-2-yl-2-oxoacetate, ethyl 5-bromothien-2-yl glyoxylate, ethyl 2-5-bromo-2-thienyl-2-oxoacetate, ethyl 5-bromothiophene-2-glyoxylate, ethyl 5-bromo-2-thienyl glyoxylate, ethyl 2-5-bromothien-2-yl glyoxalate, ethyl 5-bromothiophen-2-yl oxo acetate, 2-bromo-5-ethoxy oxo acetyl thiophene, 5-bromothiophen-2-yl oxoacetic acid ethyl ester
PubChem CID 2736376
IUPAC Name ethyl 2-(5-bromothiophen-2-yl)-2-oxoacetate
SMILES CCOC(=O)C(=O)C1=CC=C(S1)Br

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 263.11
Percent Purity 97%
Chemical Name or Material Ethyl 2-(5-bromo-2-thienyl)-2-oxoacetate
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.