accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NHFClOHO

CAS RN 220991-20-8

IUPAC Name: 2-{2-[(2-chloro-6-fluorophenyl)amino]-5-methylphenyl}acetic acid
Synonyms: lumiracoxib prexige prexige (novartis) C15H13ClFNO2
Molecular Weight (g/mol): 293.72
Molecular Formula: C15H13ClFNO2
InChi Key: KHPKQFYUPIUARC-UHFFFAOYSA-N
SMILES: CC1=CC=C(NC2=C(F)C=CC=C2Cl)C(CC(O)=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results