accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 221006-66-2

IUPAC Name: (3-phenoxyphenyl)boronic acid
Synonyms: 3-phenoxyphenylboronic acid
Molecular Weight (g/mol): 214.03
Molecular Formula: C12H11BO3
InChi Key: LOPQWMNOCSRRSR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(OC2=CC=CC=C2)=CC=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results