CAS RN 221012-82-4
IUPAC Name:
4,4'-bis(diphenylphosphanyl)-2,2',6,6'-tetramethoxy-3,3'-bipyridine
Synonyms:
4,4'-bis(diphenylphosphanyl)-2,2',6,6'-tetramethoxy-3,3'-bipyridine
Molecular Weight (g/mol):
644.65
Molecular Formula:
C38H34N2O4P2
InChi Key:
JZOSBBLJKXSBBN-UHFFFAOYSA-N
SMILES:
COC1=NC(OC)=C(C(=C1)P(C1=CC=CC=C1)C1=CC=CC=C1)C1=C(OC)N=C(OC)C=C1P(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Sigma Aldrich (4S)-(+)-4-[4-(Tert-Butyl)Phenyl]-Alpha-[(4S)-4-[4-(Tert-Butyl)Phenyl]-2-Oxazolidinylidene]-2-Oxazolineacetonitrile
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.