CAS RN 221244-14-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ PP1 Analog II, 1NM-PP1
PP1 Analog II, CAS 221244-14-0, is a cell-permeable PP1 analog that acts as a potent, reversible, selective, ATP-competitive inhibitor of mutant over wild-type kinases.