accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2ClClSNH2OO

CAS RN 22134-75-4

IUPAC Name: 4-amino-3,5-dichlorobenzene-1-sulfonamide
Synonyms: 4-amino-3,5-dichlorobenzenesulfonamide
Molecular Weight (g/mol): 241.09
Molecular Formula: C6H6Cl2N2O2S
InChi Key: DVZMRTJKNJKEGV-UHFFFAOYSA-N
SMILES: NC1=C(Cl)C=C(C=C1Cl)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results