Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3,5-Dichlorosulfanilamide 98.0+%, TCI America™
SDP

Catalog No. D04145G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D04145G 5 g
D041425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D04145G Supplier TCI America Supplier No. D04145G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 22134-75-4
Molecular Formula C6H6Cl2N2O2S
Molecular Weight (g/mol) 241.086
MDL Number MFCD00014784
InChI Key DVZMRTJKNJKEGV-UHFFFAOYSA-N
Synonym 3,5-dichlorosulfanilamide, benzenesulfonamide, 4-amino-3,5-dichloro, 4-amino-3,5-dichlorobenzene-1-sulfonamide, sulfanilamide, 3,5-dichloro, 4-amino-3,5-dichloro-benzenesulfonamide, 3,5-dichlorosulphanilamide, 1zge, acmc-1clhm, sulfanilamide,5-dichloro, aronis24244
PubChem CID 89607
IUPAC Name 4-amino-3,5-dichlorobenzenesulfonamide
SMILES C1=C(C=C(C(=C1Cl)N)Cl)S(=O)(=O)N

Specifications

Melting Point 206°C
Quantity 5 g
Formula Weight 241.09
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material 3,5-Dichlorosulfanilamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.