accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3CH3CH3OHOO

CAS RN 2215-21-6

IUPAC Name: 2-hydroxy-3,5-bis(propan-2-yl)benzoate
Synonyms: 2-hydroxy-3,5-diisopropylbenzoate
Molecular Weight (g/mol): 221.28
Molecular Formula: C13H17O3
InChi Key: XUFUYOGWFZSHGE-UHFFFAOYSA-M
SMILES: CC(C)C1=CC(C(C)C)=C(O)C(=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results