accessibility menu, dialog, popup

UserName

Viewing profile as user:
OHNN

CAS RN 2215-63-6

IUPAC Name: 1-benzyl-2,3-dihydro-1H-indazol-3-one
Synonyms: 1-benzyl-2H-indazol-3-one
Molecular Weight (g/mol): 224.26
Molecular Formula: C14H12N2O
InChi Key: SXPJFDSMKWLOAB-UHFFFAOYSA-N
SMILES: O=C1NN(CC2=CC=CC=C2)C2=CC=CC=C12
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results