Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Benzyl-1H-indazol-3-ol 98.0+%, TCI America™
SDP

Catalog No. B24115G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B24115G Supplier TCI America Supplier No. B24115G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 2215-63-6
Melting Point 167°C
Molecular Formula C14H12N2O
Quantity 5 g
MDL Number MFCD01631173
Synonym 1-benzyl-3-hydroxy-1h-indazole, 1-benzyl-1h-indazol-3-ol, 1-benzyl-1,2-dihydro-3h-indazol-3-one, 1-benzylindazol-3-ol, 1-benzyl-1,2-dihydro-indazol-3-one, 1-benzyl-2,3-dihydroindazol-3-one, 1-benzyl-3-hydroxyindazole, chembl8612, 3h-indazol-3-one, 1,2-dihydro-1-phenylmethyl, benzyl-3-hydroxy-1h-indazole
InChI Key SXPJFDSMKWLOAB-UHFFFAOYSA-N
SMILES O=C1NN(CC2=CC=CC=C2)C2=CC=CC=C12
IUPAC Name 1-benzyl-2,3-dihydro-1H-indazol-3-one
Molecular Weight (g/mol) 224.26
PubChem CID 75181
Formula Weight 224.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Benzyl-1H-indazol-3-ol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.