accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COHHONHNNONNH2

CAS RN 22150-76-1

IUPAC Name: 2-amino-6-(1,2-dihydroxypropyl)-1,4-dihydropteridin-4-one
Synonyms: biopterin
Molecular Weight (g/mol): 237.22
Molecular Formula: C9H11N5O3
InChi Key: LHQIJBMDNUYRAM-UHFFFAOYNA-N
SMILES: CC(O)C(O)C1=CN=C2NC(N)=NC(=O)C2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results