Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals L-Biopterin, 98%

Catalog No. AC461662500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461662500 250 mg
AC461660010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461662500 Supplier Thermo Scientific Chemicals Supplier No. 461662500

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22150-76-1
Molecular Formula C9H11N5O3
Molecular Weight (g/mol) 237.22
InChI Key LHQIJBMDNUYRAM-UHFFFAOYNA-N
IUPAC Name 2-amino-6-(1,2-dihydroxypropyl)-1,4-dihydropteridin-4-one
SMILES CC(O)C(O)C1=CN=C2NC(N)=NC(=O)C2=N1

Specifications

Assay Percent Range 97.5 % min. (HPLC)
Quantity 250 mg
Formula Weight 237.22
Chemical Name or Material L-Biopterin
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

If eye irritation persists: Get medical advice/attention.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

If skin irritation occurs: Get medical advice/attention.

EINECSNumber 244-807-8

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.