accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CCH3(E)

CAS RN 2216-38-8

IUPAC Name: (2E)-non-2-ene
Synonyms: (2E)-non-2-ene
Molecular Weight (g/mol): 126.24
Molecular Formula: C9H18
InChi Key: IICQZTQZQSBHBY-HWKANZROSA-N
SMILES: CCCCCC\C=C\C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results