accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3

CAS RN 2216-87-7

IUPAC Name: undecan-3-one
Synonyms: 3-undecanone
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: YNMZZHPSYMOGCI-UHFFFAOYSA-N
SMILES: CCCCCCCCC(=O)CC
Boiling Point 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)