Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Undecanone 85.0+%, TCI America™
SDP

Catalog No. U00295ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U00295ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. U00295ML Supplier TCI America Supplier No. U00295ML

Specifications

CAS 2216-87-7
Color Yellow
Boiling Point 227°C
Molecular Formula C11H22O
Quantity 5 mL
MDL Number MFCD00009572
Synonym 3-undecanone, ethyl octyl ketone, ethyl n-octyl ketone, undecan-9-one, acmc-209ft6
InChI Key YNMZZHPSYMOGCI-UHFFFAOYSA-N
SMILES CCCCCCCCC(=O)CC
IUPAC Name undecan-3-one
Molecular Weight (g/mol) 170.296
PubChem CID 75189
Formula Weight 170.30
Percent Purity ≥85.0% (GC)
Physical Form Liquid
Chemical Name or Material 3-Undecanone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.