accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2NNOHF(Z)

CAS RN 22179-78-8

IUPAC Name: (Z)-4-fluoro-N'-hydroxybenzene-1-carboximidamide
Synonyms: (Z)-4-fluoro-N'-hydroxybenzenecarboximidamide
Molecular Weight (g/mol): 154.14
Molecular Formula: C7H7FN2O
InChi Key: OSUPWUQRPLIJKX-UHFFFAOYSA-N
SMILES: N\C(=N/O)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results