Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Fluorobenzamidoxime, 97%, Thermo Scientific™

Catalog No. AAH6656714
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6656714 25 g
AAH6656706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6656714 Supplier Thermo Scientific Chemicals Supplier No. H6656714

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22179-78-8
Molecular Formula C7H7FN2O
Molecular Weight (g/mol) 154.14
MDL Number MFCD00216571,MFCD02730126
InChI Key OSUPWUQRPLIJKX-UHFFFAOYSA-N
Synonym 4-fluoro-n-hydroxybenzimidamide, 4-fluoro-n'-hydroxybenzimidamide, z-4-fluoro-n'-hydroxybenzenecarboximidamide, 4-fluorophenyl hydroxyimino methylamine, benzenecarboximidamide,4-fluoro-n-hydroxy, p-fluorobenzamidoxime, #, zyb
PubChem CID 5960802
IUPAC Name (Z)-4-fluoro-N'-hydroxybenzene-1-carboximidamide
SMILES N\C(=N/O)C1=CC=C(F)C=C1

Specifications

Melting Point 79°C to 81°C
Color White
Quantity 25 g
Formula Weight 154.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluorobenzamidoxime
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.