accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2222-33-5

IUPAC Name: tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-one
Synonyms: tricyclo[9.4.0.0³,⁸]pentadeca-1(15),3,5,7,9,11,13-heptaen-2-one
Molecular Weight (g/mol): 206.24
Molecular Formula: C15H10O
InChi Key: SNVTZAIYUGUKNI-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C=CC2=CC=CC=C12
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (1)
  • (3)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)