Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dibenzosuberenone, 97%

Catalog No. AC112531000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC112531000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC112531000 Supplier Thermo Scientific Chemicals Supplier No. 112531000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2222-33-5 | C15H10O | 206.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2222-33-5
Molecular Formula C15H10O
Molecular Weight (g/mol) 206.24
MDL Number MFCD00003588
InChI Key SNVTZAIYUGUKNI-UHFFFAOYSA-N
Synonym dibenzosuberenone, 5-dibenzosuberenone, 5h-dibenzo a,d cyclohepten-5-one, 5h-dibenzo a,d 7 annulen-5-one, dibenzo a,d cyclohepten-5-one, 2,3:6,7-dibenzotropone, 2,3:6,7-dibenzosuberen-4-on-1, 5h-dibenzo a,d cycloheptenone
PubChem CID 16679
IUPAC Name dibenzo[1,2-a
SMILES C1=CC=C2C(=C1)C=CC3=CC=CC=C3C2=O

Specifications

Melting Point 85.0°C to 91.0°C
Color White to Yellow or Beige
Loss on Drying 0.5% max. (60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Quantity 100 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,toluene,chloroform,and ethyl acetate,sparingly soluble in ethanol and methanol
Formula Weight 206.24
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Dibenzosuberenone, 97%
Show More Show Less
EINECSNumber 218-737-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.