accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOOOOOHOO

CAS RN 22224-41-5

IUPAC Name: [3,5-bis(benzoyloxy)-4-hydroxyoxolan-2-yl]methyl benzoate
Synonyms: [3,5-bis(benzoyloxy)-4-hydroxyoxolan-2-yl]methyl benzoate
Molecular Weight (g/mol): 462.45
Molecular Formula: C26H22O8
InChi Key: HUHVPBKTTFVAQF-UHFFFAOYNA-N
SMILES: OC1C(OC(=O)C2=CC=CC=C2)OC(COC(=O)C2=CC=CC=C2)C1OC(=O)C1=CC=CC=C1
Physical Form 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)