Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,3,5-Tri-O-Benzoyl-α-D-Ribofuranose, MP Biomedicals™

Catalog No. ICN15396801 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15396801 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15396801 Supplier MP Biomedicals, Inc Supplier No. 0215396801
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22224-41-5
Molecular Formula C26H22O8
Molecular Weight (g/mol) 462.454
InChI Key HUHVPBKTTFVAQF-GCUWJAMBSA-N
Synonym α-D-Ribofuranose 1, 3, 5-tribenzoate
PubChem CID 68022796
IUPAC Name [(2R,5R)-3,5-dibenzoyloxy-4-hydroxyoxolan-2-yl]methyl benzoate
SMILES C1=CC=C(C=C1)C(=O)OCC2C(C(C(O2)OC(=O)C3=CC=CC=C3)O)OC(=O)C4=CC=CC=C4

Specifications

Color White
Quantity 1 g
Formula Weight 462.4
Physical Form Powder
Chemical Name or Material 1, 3, 5-Tri-O-Benzoyl-α-D-Ribofuranose
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.