accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COPOHNCH3CH3OSCH3H3C

CAS RN 22224-92-6

IUPAC Name: {ethoxy[3-methyl-4-(methylsulfanyl)phenoxy]phosphoryl}(propan-2-yl)amine
Synonyms: fenamiphos caswell no. 453A nemacur methaphenamiphos nemacur P epa pesticide chemical code 100601 phenamiphos
Molecular Weight (g/mol): 303.36
Molecular Formula: C13H22NO3PS
InChi Key: ZCJPOPBZHLUFHF-UHFFFAOYNA-N
SMILES: CCOP(=O)(NC(C)C)OC1=CC=C(SC)C(C)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results