accessibility menu, dialog, popup

UserName

Viewing profile as user:
OFFH2N

CAS RN 22236-08-4

IUPAC Name: 3-(difluoromethoxy)aniline
Synonyms: 3-(difluoromethoxy)aniline
Molecular Weight (g/mol): 159.14
Molecular Formula: C7H7F2NO
InChi Key: RSOFZRXRIPVBBM-UHFFFAOYSA-N
SMILES: NC1=CC(OC(F)F)=CC=C1
Boiling Point 
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)