Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-(Difluoromethoxy)aniline 98.0+%, TCI America™
SDP

Catalog No. D34565G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D34565G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D34565G Supplier TCI America Supplier No. D34565G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 22236-08-4
Molecular Formula C7H7F2NO
Molecular Weight (g/mol) 159.14
MDL Number MFCD00236221
InChI Key RSOFZRXRIPVBBM-UHFFFAOYSA-N
Synonym 3-difluoromethoxy aniline, m-difluoromethoxyaniline, benzenamine, 3-difluoromethoxy, 3-amino-alpha,alpha-difluoroanisole, 3-difluoromethoxy phenylamine, pubchem8466, acmc-1ckmq, a,a-difluoro-m-anisidine, 3-difluoromethoxyphenylamine
PubChem CID 2774110
IUPAC Name 3-(difluoromethoxy)aniline
SMILES NC1=CC(OC(F)F)=CC=C1

Specifications

Color Yellow
Boiling Point 92°C
Quantity 5 g
Formula Weight 159.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 3-(Difluoromethoxy)aniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.