accessibility menu, dialog, popup

UserName

Viewing profile as user:
OO(R)NH2(S)(R)

CAS RN 22255-16-9

IUPAC Name: (1R,2R,5S)-3-azabicyclo[3.1.0]hexan-3-ium-2-carboxylate
Synonyms: (1R,2R,5S)-3-azabicyclo[3.1.0]hexan-3-ium-2-carboxylate
Molecular Weight (g/mol): 127.14
Molecular Formula: C6H9NO2
InChi Key: JBDOTWVUXVXVDR-UOWFLXDJSA-N
SMILES: [O-]C(=O)[C@@H]1[NH2+]C[C@H]2C[C@@H]12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results