accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 22264-50-2

IUPAC Name: 1-aminocyclobutane-1-carboxylic acid
Synonyms: 1-aminocyclobutane-1-carboxylic acid
Molecular Weight (g/mol): 115.13
Molecular Formula: C5H9NO2
InChi Key: FVTVMQPGKVHSEY-UHFFFAOYSA-N
SMILES: NC1(CCC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)