Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Aminocyclobutanecarboxylic acid, 95%

Catalog No. AAH6445906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6445906 5 g
AAH6445903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6445906 Supplier Thermo Scientific Chemicals Supplier No. H6445906
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22264-50-2 | C5H9NO2 | 115.132 g/mol

ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
ACBC is an NMDA receptor antagonist acting at the glycine site. It acts as an agonist being an analog of glycine at the NMDA-glycine receptor site. This affects the signal transmission in the CNS.

Solubility
Soluble to 100 mM in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22264-50-2
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00661068
InChI Key FVTVMQPGKVHSEY-UHFFFAOYSA-N
Synonym 1-aminocyclobutanecarboxylic acid, acbc, 1-amino-cyclobutanecarboxylic acid, 1-aminocyclobutane carboxylic acid, 1-amino-1-cyclobutanecarboxylic acid, cyclobutanecarboxylic acid, 1-amino, 1-aminocyclobutanecarboxlic acid, 1-amino-1-carboxycyclobutane, cyclovaline
PubChem CID 89643
IUPAC Name 1-aminocyclobutane-1-carboxylic acid
SMILES C1CC(C1)(C(=O)O)N

Specifications

Melting Point 261°C (decomposition)
Color White
Quantity 5 g
Beilstein 2802253
Solubility Information Soluble to 100 mM in water.
Formula Weight 115.13
Percent Purity 95%
Physical Form Powder
Chemical Name or Material 1-Aminocyclobutanecarboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
RTECSNumber GU1336000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.