CAS RN 2227-13-6
IUPAC Name:
1,2,4-trichloro-5-[(4-chlorophenyl)sulfanyl]benzene
Synonyms:
tetrasul
Molecular Weight (g/mol):
324.04
Molecular Formula:
C12H6Cl4S
InChi Key:
QUWSDLYBOVGOCW-UHFFFAOYSA-N
SMILES:
ClC1=CC=C(SC2=CC(Cl)=C(Cl)C=C2Cl)C=C1