Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PESTANAL™ Tetrasul Analytical Standard, MilliporeSigma™ Supelco™

Catalog No. 111022467 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-2467 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-102-2467 Supplier MilliporeSigma Supelco Supplier No. 3656850MG
Add to Cart
Add to Cart

Specifications

CAS 2227-13-6
Molecular Formula C12H6Cl4S
Linear Formula C12H6Cl4S
Quantity 50 mg
MDL Number MFCD00018649
UN Number NONH for all modes of transport
Synonym 2,4,4',5-Tetrachlorodiphenyl sulfide
Formula Weight 324.05
Grade Analytical
Physical Form Neat

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.