accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2227-70-5

IUPAC Name: 2-(3-methylphenyl)-4-phenyl-1,3-thiazole
Synonyms: 2-(3-methylphenyl)-4-phenyl-1,3-thiazole
Molecular Weight (g/mol): 251.35
Molecular Formula: C16H13NS
InChi Key: QTYZJFDPFQNIML-UHFFFAOYSA-N
SMILES: CC1=CC=CC(=C1)C1=NC(=CS1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results