CAS RN 22282-58-2
IUPAC Name:
2-iodo-3-methylpyridine
Synonyms:
2-iodo-3-methylpyridine
Molecular Weight (g/mol):
219.03
Molecular Formula:
C6H6IN
InChi Key:
PTBOOHAPESUXGV-UHFFFAOYSA-N
SMILES:
CC1=CC=CN=C1I
1
–
1
of
1
results
1
Promotions Available
2-Iodo-3-methylpyridine, 97%
2-Iodo-3-methylpyridine, 97%
CAS: 22282-58-2 Molecular Formula: C6H6IN Molecular Weight (g/mol): 219.03 MDL Number: MFCD04039350 InChI Key: PTBOOHAPESUXGV-UHFFFAOYSA-N PubChem CID: 339045 IUPAC Name: 2-iodo-3-methylpyridine SMILES: CC1=CC=CN=C1I