Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Iodo-3-methylpyridine, 97%

Catalog No. AAL2004706
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL2004706 5 g
AAL2004703 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL2004706 Supplier Thermo Scientific Chemicals Supplier No. L2004706
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 22282-58-2 | C6H6IN | 219.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22282-58-2
Molecular Formula C6H6IN
Molecular Weight (g/mol) 219.03
MDL Number MFCD04039350
InChI Key PTBOOHAPESUXGV-UHFFFAOYSA-N
PubChem CID 339045
IUPAC Name 2-iodo-3-methylpyridine
SMILES CC1=CC=CN=C1I

Specifications

Boiling Point 80°C (0.3 mmHg)
Refractive Index 1.625
Quantity 5 g
Beilstein 5328169
Sensitivity Light sensitive
Synonym 2-iodo-3-methyl-pyridine, pyridine, 2-iodo-3-methyl, pubchem14399, acmc-1cgxh, pyridine,2-iodo-3-methyl, 2-iodanyl-3-methyl-pyridine, 2-iodo-3-methylpyridine, 2-iodo-3-methyl pyridine
Formula Weight 219.03
Percent Purity 97%
Chemical Name or Material 2-Iodo-3-methylpyridine
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.